Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lancl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133737.2 | NP_598498.1 | 9 | 1970 | ||
GenBank mRNA | AB035967.1 | - | 9 | 1789 | ||
AK005632.1 | - | 9 | 1338 |