Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 5 | 2710 | ||
GenBank mRNA | AK010163.1 | - | 3 | 2203 | ||
AK150361.1 | - | 5 | 2658 | |||
AK150365.1 | - | 6 | 2738 | |||
AK150630.1 | - | 1 | 1860 | |||
AK151044.1 | - | 5 | 2677 | |||
AK152423.1 | - | 5 | 2623 | |||
AK153122.1 | - | 5 | 2623 | |||
AK153378.1 | - | 6 | 2738 | |||
AK155767.1 | - | 5 | 2678 | |||
AK157001.1 | - | 6 | 2635 | |||
AK162610.1 | - | 5 | 2704 | |||
AK165671.1 | - | 5 | 2672 | |||
AK166444.1 | - | 5 | 2684 | |||
AK170823.1 | - | 5 | 2669 | |||
AK172352.1 | - | 5 | 2640 | |||
AK172553.1 | - | 5 | 2639 | |||
AK172701.1 | - | 5 | 2640 | |||
BC010292.1 | - | 5 | 2661 | 3153174 | ||
BC086319.1 | - | 5 | 2667 | 5293409 |