Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Insig2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271531.1 | NP_001258460.1 | 6 | 983 | ||
NM_001271532.1 | NP_001258461.1 | 5 | 850 | |||
NM_133748.2 | NP_598509.1 | 6 | 1226 | |||
NM_178082.3 | NP_835183.1 | 6 | 990 | |||
GenBank mRNA | AF527631.1 | - | 6 | 960 | ||
AK040703.1 | - | 6 | 981 | |||
AK050394.1 | - | 6 | 984 | |||
AK152076.1 | - | 6 | 956 | |||
AK154543.1 | - | 6 | 989 | |||
AK161256.1 | - | 6 | 994 | |||
AK165634.1 | - | 6 | 995 | |||
AK217506.1 | - | 1 | 72 | |||
BC015288.1 | - | 6 | 940 | 4240711 | ||
BC023067.1 | - | 6 | 964 | 5357599 | ||
BC023874.1 | - | 5 | 882 | 5355039 | ||
BC024411.1 | - | 6 | 978 | 4971297 |