Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncapg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133762.3 | NP_598523.3 | 25, 26 | 3339 | ||
GenBank mRNA | AY455829.1 | - | 19, 20 | 2450 | ||
BC008539.1 | - | 14, 15 | 2019 | |||
BC076631.1 | - | 24, 25 | 3149 | 30637859 |