Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133805.3 | NP_598566.3 | 3 | 309 | ||
GenBank mRNA | AF482000.1 | - | 3 | 770 | ||
AK079111.1 | - | 3 | 243 | |||
AK079522.1 | - | 3 | 257 | |||
AK088323.1 | - | 3 | 229 | |||
AK090013.1 | - | 3 | 238 | |||
AK142989.1 | - | 2 | 1278 | |||
AK145899.1 | - | 3 | 236 | |||
AK162502.1 | - | 3 | 244 | |||
AK195692.1 | - | 2 | 84 | |||
BC017690.1 | - | 3 | 277 | 4238947 | ||
BC021488.1 | - | 3 | 268 | 5344864 | ||
BC024421.1 | - | 3 | 262 | 4987584 |