Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133819.3 | NP_598580.1 | 1 | 753 | ||
GenBank mRNA | AK049028.1 | - | 1 | 457 | ||
AK082957.1 | - | 1 | 589 | |||
AK086606.1 | - | 1 | 385 | |||
AK144678.1 | - | 1 | 506 | |||
AK146089.1 | - | 1 | 491 | |||
AK167565.1 | - | 1 | 576 | |||
AK170862.1 | - | 1 | 583 | |||
BC058078.1 | - | 1 | 536 |