Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133859.2 | NP_598620.2 | 2 | 209 | ||
GenBank mRNA | AK002518.1 | - | 2 | 173 | ||
AK076084.1 | - | 2 | 175 | |||
AK131879.1 | - | 173 | ||||
AK143728.1 | - | 761 | ||||
BC005485.1 | - | 2 | 156 | 3485412 | ||
BY059553.1 | - | 2 | 210 |