Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133879.2 | NP_598640.2 | 2 | 422 | ||
GenBank mRNA | AK027912.1 | - | 2 | 423 | ||
AM161454.1 | - | 2 | 370 | |||
BC005515.1 | - | 2 | 433 | 3493611 | ||
BY269141.1 | - | 2 | 423 |