Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 2 | 263 | ||
GenBank mRNA | AB019211.1 | - | 2 | 206 | ||
AK076390.1 | - | 248 | ||||
AK077678.1 | - | 2 | 264 | |||
AK078948.1 | - | 2 | 223 | |||
AK132618.1 | - | 2 | 227 | |||
AK146544.1 | - | 2 | 268 | |||
AK146735.1 | - | 2 | 255 | |||
AK167087.1 | - | 2 | 177 | |||
AK167804.1 | - | 2 | 227 | |||
AK168753.1 | - | 2 | 221 | |||
AK170203.1 | - | 2 | 300 | |||
AK187081.1 | - | 1 | 67 | |||
BC010669.1 | - | 2 | 250 | 2645010 |