Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133889.2 | NP_598650.2 | 3, 4 | 201 | ||
GenBank mRNA | AK037201.1 | - | 3, 4 | 197 | ||
AK144053.1 | - | 3, 4 | 196 | |||
BC021480.1 | - | 2, 3 | 181 | |||
BC049111.1 | - | 3, 4 | 201 | 6415574 |