Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4bp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133898.4 | NP_598659.3 | 5 | 806 | ||
GenBank mRNA | AK046116.1 | - | 5 | 783 | ||
AK131647.1 | - | 5 | 798 | |||
BC019754.1 | - | 4 | 768 | 4007588 |