Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spon2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133903.3 | NP_598664.3 | 3 | 639 | ||
GenBank mRNA | AK028987.1 | - | 3 | 584 | ||
AK042840.1 | - | 3 | 587 | |||
AK131131.1 | - | 2 | 222 | |||
AY457639.1 | - | 3 | 374 | |||
BC017616.1 | - | 3 | 572 | 4923292 |