Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133905.2 | NP_598666.1 | 3, 4 | 455 | ||
GenBank mRNA | AK146086.1 | - | 3, 4 | 439 | ||
AM419011.1 | - | 2, 3 | 228 | |||
BC016629.1 | - | 3, 4 | 455 | 3595512 |