Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zkscan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029869.1 | NP_084145.1 | 2 | 318 | ||
NM_133906.4 | NP_598667.2 | 2 | 324 | |||
GenBank mRNA | AK020039.1 | - | 2 | 300 | ||
AK028932.1 | - | 2 | 255 | |||
AK031759.1 | - | 2 | 305 | |||
AK036807.1 | - | 2 | 307 | |||
AK054323.1 | - | 2 | 295 | |||
AK131148.1 | - | 1 | 463 | |||
AK135999.1 | - | 2 | 274 | |||
AK154831.1 | - | 2 | 304 | |||
BC052441.1 | - | 2 | 282 | 6406518 | ||
BY282109.1 | - | 2 | 325 |