Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Emilin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133918.2 | NP_598679.1 | 1 | 179 | ||
GenBank mRNA | AK020015.1 | - | 1 | 226 | ||
AK029337.1 | - | 1 | 150 | |||
AK052368.1 | - | 1 | 260 | |||
AK088995.1 | - | 1 | 242 | |||
AK138020.1 | - | 1 | 201 | |||
AK155486.1 | - | 1 | 481 | |||
BC005481.1 | - | 1 | 179 | 3485057 |