Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164724.1 | NP_001158196.1 | 4, 5 | 422 | ||
NM_133775.2 | NP_598536.2 | 4, 5 | 401 | |||
GenBank mRNA | AK020353.1 | - | 4, 5 | 402 | ||
AK075849.1 | - | 4, 5 | 423 | |||
AK163464.1 | - | 4, 5 | 402 | |||
AY905582.1 | - | 3, 4 | 342 | |||
BC003847.1 | - | 4, 5 | 378 | 3593927 | ||
HQ176370.1 | - | 1, 2 | 42 |