Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133781.4 | NP_598542.3 | 2, 3 | 540 | ||
GenBank mRNA | AK144665.1 | - | 3, 4 | 373 | ||
BC020041.1 | - | 2, 3 | 202 | 3990872 | ||
S51858.1 | - | 2, 3 | 277 |