Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133798.3 | NP_598559.2 | 3, 4 | 915 | ||
GenBank mRNA | AK146901.1 | - | 3, 4 | 796 | ||
AK147922.1 | - | 3, 4 | 776 | |||
AK158201.1 | - | 3, 4 | 806 | |||
AK162206.1 | - | 2, 3 | 683 | |||
AK164329.1 | - | 3, 4 | 791 | |||
AK166390.1 | - | 3, 4 | 797 | |||
AK167692.1 | - | 3, 4 | 916 | |||
BC018508.1 | - | 2, 3 | 458 | 3156447 |