Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133807.1 | NP_598568.1 | 4 | 567 | ||
GenBank mRNA | AK053450.1 | - | 4 | 594 | ||
AK159374.1 | - | 4 | 623 | |||
AK159552.1 | - | 4 | 617 | |||
AK166576.1 | - | 4 | 616 | |||
AK167604.1 | - | 4 | 558 | |||
AK169250.1 | - | 4 | 542 | |||
AK169390.1 | - | 4 | 566 | |||
AK196953.1 | - | 140 | ||||
BC006877.1 | - | 4 | 567 | 2647513 | ||
BC066172.1 | - | 4 | 561 | 30429192 |