Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133809.1 | NP_598570.1 | 3 | 227 | ||
GenBank mRNA | AK129011.1 | - | 240 | |||
AK133074.1 | - | 335 | ||||
AK171812.1 | - | 3 | 351 | |||
BC014683.1 | - | 3 | 227 | 4210278 |