Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp1r15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133819.3 | NP_598580.1 | 1 | 1054 | ||
GenBank mRNA | AK049028.1 | - | 1 | 758 | ||
AK082957.1 | - | 1 | 890 | |||
AK086606.1 | - | 1 | 686 | |||
AK144678.1 | - | 1 | 807 | |||
AK146089.1 | - | 1 | 792 | |||
AK167565.1 | - | 1 | 877 | |||
AK170862.1 | - | 1 | 884 | |||
BC058078.1 | - | 1 | 837 |