Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025312.1 | NP_001020483.1 | 2 | 621 | ||
NM_133865.2 | NP_598626.2 | 3 | 787 | |||
GenBank mRNA | AK046571.1 | - | 3 | 790 | ||
AK051104.1 | - | 3 | 742 | |||
AK084347.1 | - | 2 | 616 | |||
AK134324.1 | - | 3 | 547 | |||
AK137495.1 | - | 3 | 344 | |||
AK138568.1 | - | 3 | 845 | |||
AK154124.1 | - | 3 | 689 | |||
AK163827.1 | - | 2 | 1388 | |||
BC011094.1 | - | 2 | 246 | 3493038 | ||
BC067017.1 | - | 2 | 530 | 6827667 | ||
BC125277.1 | - | 2 | 242 | |||
BC144725.1 | - | 2 | 242 |