Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bsdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133889.2 | NP_598650.2 | 4 | 229 | ||
GenBank mRNA | AK037201.1 | - | 4 | 225 | ||
AK084857.1 | - | 3 | 1701 | |||
AK144053.1 | - | 4 | 224 | |||
BC021480.1 | - | 3 | 209 | |||
BC049111.1 | - | 4 | 229 | 6415574 |