Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133900.4 | NP_598661.1 | 5 | 794 | ||
GenBank mRNA | AK088865.1 | - | 5 | 746 | ||
AK168048.1 | - | 3 | 402 | |||
AK213907.1 | - | 2 | 43 | |||
BC002251.1 | - | 3 | 365 | 3493291 | ||
BQ177662.1 | - | 3 | 360 |