Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube3c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133907.3 | NP_598668.1 | 2 | 356 | ||
GenBank mRNA | AK029973.1 | - | 2 | 166 | ||
AK045211.1 | - | 2 | 153 | |||
AK046056.1 | - | 2 | 357 | |||
AK122187.1 | - | 2 | 291 | |||
AK140491.1 | - | 2 | 113 | |||
BC059245.1 | - | 2 | 172 | |||
BC120731.1 | - | 2 | 103 | 40129654 | ||
BC137626.1 | - | 2 | 103 | 8860645 |