Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000547.5 | NP_000538.3 | 4 | 295 | ||
NM_001206744.1 | NP_001193673.1 | 4 | 290 | |||
NM_001206745.1 | NP_001193674.1 | 4 | 290 | |||
NM_175719.3 | NP_783650.1 | 4 | 295 | |||
NM_175721.3 | NP_783652.1 | 3 | 205 | |||
NM_175722.3 | NP_783653.1 | 3 | 205 | |||
XM_011510379.2 | XP_011508681.1 | 3 | 205 | |||
XM_011510380.2 | XP_011508682.1 | 3 | 205 | |||
XM_011510381.2 | XP_011508683.1 | 3 | 205 | |||
GenBank mRNA | AF439430.1 | - | 3 | 204 | ||
AF533528.1 | - | 2 | 123 | |||
DA958481.1 | - | 3 | 231 | |||
J02969.1 | - | 4 | 276 | |||
J02970.1 | - | 4 | 276 | |||
X17358.1 | - | 4 | 269 | |||
Y00406.1 | - | 4 | 244 |