Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164724.1 | NP_001158196.1 | 8 | 2281 | ||
NM_133775.2 | NP_598536.2 | 8 | 2260 | |||
GenBank mRNA | AK075849.1 | - | 8 | 2283 | ||
AK163464.1 | - | 8 | 2261 | |||
BC003847.1 | - | 8 | 2237 | 3593927 | ||
CJ191135.1 | - | 1 | 114 |