Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133777.2 | NP_598538.1 | 4 | 787 | ||
GenBank mRNA | AK003078.1 | - | 4 | 801 | ||
AK017648.1 | - | 4 | 803 | |||
AK146276.1 | - | 4 | 799 | |||
AK154026.1 | - | 4 | 800 | |||
AK154067.1 | - | 4 | 803 | |||
BC012255.1 | - | 4 | 787 | 3581845 | ||
BC030171.1 | - | 4 | 781 | 5037250 | ||
BC083323.1 | - | 4 | 769 | 5715696 |