Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Exd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133798.3 | NP_598559.2 | 7 | 1552 | ||
GenBank mRNA | AK146901.1 | - | 7 | 1433 | ||
AK147922.1 | - | 7 | 1413 | |||
AK158201.1 | - | 7 | 1443 | |||
AK162206.1 | - | 6 | 1320 | |||
AK164329.1 | - | 7 | 1428 | |||
AK167692.1 | - | 7 | 1553 | |||
BC018508.1 | - | 6 | 1095 | 3156447 |