Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133805.3 | NP_598566.3 | 8 | 1542 | ||
GenBank mRNA | AF482000.1 | - | 8 | 2003 | ||
AK079111.1 | - | 8 | 1476 | |||
AK079522.1 | - | 8 | 1490 | |||
AK088323.1 | - | 8 | 1462 | |||
AK090013.1 | - | 8 | 1471 | |||
AK142989.1 | - | 7 | 2511 | |||
AK145899.1 | - | 8 | 1469 | |||
AK162502.1 | - | 8 | 1477 | |||
AK207412.1 | - | 1 | 309 | |||
AK212344.1 | - | 1 | 307 | |||
AK214410.1 | - | 1 | 306 | |||
AK215258.1 | - | 1 | 309 | |||
AK218596.1 | - | 1 | 307 | |||
BC017690.1 | - | 8 | 1509 | 4238947 | ||
BC021488.1 | - | 8 | 1500 | 5344864 | ||
BC024421.1 | - | 8 | 1494 | 4987584 |