Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdlbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301364.1 | NP_001288293.1 | 28 | 6026 | ||
NM_133808.5 | NP_598569.1 | 28 | 6109 | |||
GenBank mRNA | AK003484.1 | - | 1 | 557 | ||
BC027788.1 | - | 28 | 6004 | 5346979 | ||
BC046545.1 | - | 8 | 2929 | |||
CK638858.1 | - | 1 | 435 |