Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kmo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133809.1 | NP_598570.1 | 15 | 1781 | ||
GenBank mRNA | AK129011.1 | - | 1692 | |||
AK171812.1 | - | 15 | 1909 | |||
BC014683.1 | - | 15 | 1781 | 4210278 | ||
BC024455.1 | - | 4 | 733 |