Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3bp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133816.2 | NP_598577.1 | 4 | 2215 | ||
GenBank mRNA | AK028826.1 | - | 4 | 2195 | ||
AK032634.1 | - | 4 | 2256 | |||
AK041504.1 | - | 4 | 2263 | |||
AK043259.1 | - | 1 | 441 | |||
BC012284.1 | - | 4 | 2215 | 3978494 | ||
BC028911.1 | - | 1 | 1450 | |||
BC034733.1 | - | 3 | 2051 | 4947387 |