Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133887.4 | NP_598648.1 | 4 | 470 | ||
GenBank mRNA | AB019211.1 | - | 4 | 413 | ||
AK076390.1 | - | 455 | ||||
AK077678.1 | - | 4 | 471 | |||
AK078948.1 | - | 4 | 430 | |||
AK132618.1 | - | 4 | 433 | |||
AK146544.1 | - | 4 | 475 | |||
AK146735.1 | - | 4 | 462 | |||
AK167087.1 | - | 4 | 384 | |||
AK167804.1 | - | 4 | 434 | |||
AK168753.1 | - | 4 | 428 | |||
AK170203.1 | - | 4 | 507 | |||
BC010669.1 | - | 4 | 457 | 2645010 |