Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133900.4 | NP_598661.1 | 7 | 1109 | ||
GenBank mRNA | AA003039.1 | - | 1 | 54 | ||
AK088865.1 | - | 7 | 1061 | |||
AK168048.1 | - | 5 | 717 | |||
AK217223.1 | - | 2 | 130 | |||
BC002251.1 | - | 5 | 680 | 3493291 |