Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Papd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133905.2 | NP_598666.1 | 7 | 919 | ||
GenBank mRNA | AK033141.1 | - | 4 | 254 | ||
BC016629.1 | - | 7 | 919 | 3595512 |