Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tada3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133932.2 | NP_598693.1 | 2 | 333 | ||
GenBank mRNA | AK078906.1 | - | 2 | 653 | ||
BC023879.1 | - | 3 | 396 | |||
BC026630.1 | - | 2 | 333 | 4983657 | ||
BC032195.1 | - | 3 | 254 | 5323331 |