Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 4 | 835 | ||
GenBank mRNA | AK041905.1 | - | 4 | 704 | ||
AK089681.1 | - | 680 | ||||
AK090333.1 | - | 4 | 680 | |||
AK157968.1 | - | 4 | 688 | |||
AK170639.1 | - | 4 | 688 | |||
AK170923.1 | - | 4 | 658 | |||
AK172462.1 | - | 4 | 669 | |||
AK201914.1 | - | 2 | 193 | |||
AK218562.1 | - | 2 | 193 | |||
AY166673.1 | - | 2 | 353 | |||
BC013467.1 | - | 3 | 386 | 4218025 | ||
BC075666.1 | - | 4 | 835 | 30605163 |