Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133984.2 | NP_598745.2 | 2 | 355 | ||
GenBank mRNA | AK054445.1 | - | 2 | 306 | ||
AK088071.1 | - | 2 | 323 | |||
BC011431.1 | - | 1 | 300 | 3981356 | ||
BC024428.1 | - | 1 | 160 | 4971671 |