Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rdh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134006.4 | NP_598767.1 | 3 | 567 | ||
GenBank mRNA | AF013288.1 | - | 3 | 438 | ||
AF033195.1 | - | 3 | 505 | |||
AF033196.2 | - | 3 | 533 | |||
AK033063.1 | - | 3 | 549 | |||
AK135139.1 | - | 3 | 568 | |||
BC021372.1 | - | 3 | 480 | 5040142 |