Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035226.1 | NP_001030303.1 | 16 | 1805 | ||
NM_134014.3 | NP_598775.2 | 16 | 2043 | |||
GenBank mRNA | AK136538.1 | - | 9 | 1206 | ||
BC012276.1 | - | 3 | 267 | 3156478 | ||
BC025628.1 | - | 2 | 227 | 5321933 | ||
BC029714.1 | - | 14 | 1566 | |||
BC034710.1 | - | 15 | 1800 | |||
BC062912.1 | - | 16 | 1805 | 6821729 | ||
BU052960.1 | - | 2 | 148 | |||
CF538023.1 | - | 5 | 646 |