Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipas39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142580.1 | NP_001136052.1 | 3 | 242 | ||
NM_001142581.1 | NP_001136053.1 | 3 | 242 | |||
NM_134044.3 | NP_598805.2 | 3 | 256 | |||
GenBank mRNA | AK034304.1 | - | 1 | 1453 | ||
AK038165.1 | - | 3 | 210 | |||
AK046322.1 | - | 3 | 213 | |||
AK049643.1 | - | 3 | 189 | |||
AK079055.1 | - | 3 | 189 | |||
AK082261.1 | - | 3 | 170 | |||
AK145043.1 | - | 225 | ||||
BB610654.1 | - | 3 | 243 | |||
BC016646.1 | - | 3 | 165 | 2651233 | ||
BC023716.1 | - | 3 | 267 | 5346489 |