Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pelo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134058.3 | NP_598819.2 | 1 | 753 | ||
GenBank mRNA | AF148638.1 | - | 814 | |||
AK146382.1 | - | 1 | 783 | |||
AK168228.1 | - | 1 | 783 | |||
AK170635.1 | - | 1 | 753 | |||
BC050209.1 | - | 1 | 666 | 6431710 | ||
BC057160.1 | - | 1 | 554 | 5349483 |