Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc17a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164743.1 | NP_001158215.1 | 3 | 486 | ||
NM_134069.3 | NP_598830.2 | 4 | 610 | |||
GenBank mRNA | AA986956.1 | - | 2 | 137 | ||
AK128896.1 | - | 4 | 594 | |||
AK143896.1 | - | 4 | 582 | |||
AK165505.1 | - | 4 | 455 | |||
BC018252.1 | - | 4 | 539 | 4240254 | ||
BC024459.1 | - | 4 | 566 | 4975341 | ||
BC034179.1 | - | 4 | 430 | 4972500 | ||
BC034183.1 | - | 4 | 415 | 4973620 |