Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000587.2 | NP_000578.2 | 4, 5 | 402 | ||
GenBank mRNA | AK290349.1 | - | 4, 5 | 412 | ||
AK304848.1 | - | 4, 5 | 355 | |||
AK312737.1 | - | 4, 5 | 336 | |||
BC025402.1 | - | 4, 5 | 481 | |||
BC063851.1 | - | 4, 5 | 383 | 6184213 | ||
J03507.1 | - | 3, 4 | 288 |