Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133967.3 | NP_598728.1 | 4 | 866 | ||
GenBank mRNA | AK041905.1 | - | 4 | 735 | ||
AK089681.1 | - | 711 | ||||
AK090333.1 | - | 4 | 712 | |||
AK157968.1 | - | 4 | 719 | |||
AK170639.1 | - | 4 | 719 | |||
AK170923.1 | - | 4 | 689 | |||
AK172462.1 | - | 4 | 700 | |||
AK201914.1 | - | 2 | 224 | |||
AK218562.1 | - | 2 | 224 | |||
AY166673.1 | - | 2 | 384 | |||
BC013467.1 | - | 3 | 417 | 4218025 | ||
BC075666.1 | - | 4 | 866 | 30605163 |