Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167967.2 | NP_001161439.1 | 4 | 734 | ||
NM_001281974.1 | NP_001268903.1 | 5 | 846 | |||
NM_001281975.1 | NP_001268904.1 | 5 | 896 | |||
NM_133971.3 | NP_598732.2 | 4 | 734 | |||
GenBank mRNA | AK014768.1 | - | 5 | 725 | ||
AK031808.1 | - | 4 | 732 | |||
AK033623.1 | - | 4 | 732 | |||
AK038156.1 | - | 4 | 732 | |||
AK039879.1 | - | 2 | 179 | |||
AK042500.1 | - | 4 | 732 | |||
AK050983.1 | - | 4 | 735 | |||
AK081754.1 | - | 5 | 894 | |||
AK082875.1 | - | 4 | 731 | |||
AK084456.1 | - | 5 | 844 | |||
AK086012.1 | - | 2 | 1089 | |||
AK088371.1 | - | 4 | 684 | |||
BC002198.1 | - | 4 | 666 | 3489133 | ||
BC033266.1 | - | 1 | 29 |