Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cog4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133973.2 | NP_598734.1 | 1, 2 | 191 | ||
GenBank mRNA | AK156466.1 | - | 1, 2 | 182 | ||
AK172465.1 | - | 1, 2 | 182 | |||
BC023120.1 | - | 1, 2 | 168 | 4161858 | ||
CN843612.1 | - | 1, 2 | 210 |