Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdcp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133974.3 | NP_598735.2 | 3 | 650 | ||
GenBank mRNA | AK035674.1 | - | 3 | 612 | ||
AK087108.1 | - | 3 | 548 | |||
BC018410.1 | - | 3 | 661 | |||
BC034137.1 | - | 1 | 58 | |||
BC059937.1 | - | 3 | 557 | |||
BC085253.1 | - | 3 | 650 | 5347122 |