Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trip12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133975.4 | NP_598736.4 | 19, 20 | 3005 | ||
GenBank mRNA | AK033327.1 | - | 2, 3 | 320 | ||
AK158807.1 | - | 19, 20 | 3005 | |||
BC034113.1 | - | 3, 4 | 411 | |||
BC044869.1 | - | 16, 17 | 1963 |